2,7-Dinitrofluorene, Sigma-Aldrich®
Catalog # SIALD196401-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:256.22 g/mol
Melting Pt:340 °C
EINECS:226-457-8
CAS Number:5405-53-8
Formula:C₁₃H₈N₂O₄
MDL Number:MFCD00001121