Order Entry
Export
ContactUsLinkComponent
1,8-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
1,8-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIALD108103-500G
Supplier:  Merck
CAS Number:  
1,8-Dihydroxy-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIALD108103-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :240.22 g/mol
Merck Index:13,02841
Melting Pt:190…193 °C
Density:1.537 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Flash Pt:>200 °C
EINECS:204-173-5
CAS Number:117-10-2
Formula:C₁₄H₈O₄
MDL Number:MFCD00001211