Order Entry
Export
ContactUsLinkComponent
 
 
 
1,8-Dihydroxy-9,10-anthraquinone 95%
 
undefined
 
 
1,8-Dihydroxy-9,10-anthraquinone 95%

 

  • Appearance:
    Yellow-green to orange to brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    190 to 195 °C
  • HPLC:
    ≥94 %