0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1-Hydroxy-9-fluorenone
1-Hydroxy-9-fluorenone
1-Hydroxy-9-fluorenone
1-Hydroxy-9-fluorenone
Supplier:
CHEMPUR
Certificates
Expand image
1-Hydroxy-9-fluorenone
Supplier:
CHEMPUR
About this item
CAS Number:
6344-60-1
:
Formula:
C₁₃H₈O₂
:
196.21 g/mol
Boiling Pt:
353 °C (1013 hPa)
Melting Pt:
115 °C
Density:
1.39 g/cm³ (20 °C)
EINECS:
228-746-4