2,7-Dinitro-9-fluorenone, Sigma-Aldrich®
Catalog # SIAL107107-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:270.2 g/mol
Melting Pt:296 °C
:M-Clarity™ quality level = MQ100
EINECS:250-695-1
CAS Number:31551-45-8
Formula:C₁₃H₆N₂O₅
MDL Number:MFCD00001153