1,5-Dichloro-9,10-anthraquinone, Sigma-Aldrich®
Catalog # SIALD56209-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:277.11 g/mol
Boiling Pt:401 °C (1013 hPa)
Melting Pt:245…250 °C
Density:1.63 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:201-424-0
CAS Number:82-46-2
Formula:C₁₄H₆Cl₂O₂
MDL Number:MFCD00001190