Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,7-Dibromo-9-fluorenone
2,7-Dibromo-9-fluorenone 96%
2,7-Dibromo-9-fluorenone 96%
Irritant
2,7-Dibromo-9-fluorenone 96%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
2,7-Dibromo-9-fluorenone 96%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
14348-75-5
:
Formula:
C₁₃H₆Br₂O
MDL Number:
MFCD00010790
:
338 g/mol
Melting Pt:
195…196 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH25919
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation