Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dibenz[a,c]anthracene

Glass bottle for solids, 100 mg
Catalog # PROOBCR-094
Dibenz[a,c]anthracene
Catalog # PROOBCR-094
CAS Number:  215-58-7
Dibenz[a,c]anthracene

Glass bottle for solids, 100 mg

CAS Number:  215-58-7
Price
643,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  1,2,3,4-Dibenzanthracene
  • CAS Number:215-58-7
  • Formula:C₂₂H₁₄
  • Molecular Weight:278.35 g/mol
  • Boiling Pt:518 °C (1013 hPa)
  • Melting Pt:202…207 °C
  • Storage Temperature:Ambient
  • EINECS:205-920-8
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications