

Piperazine
Catalog # MOLEM29428370
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:86.14 g/mol
Merck Index:13,07545
Boiling Pt:146 °C (1013 hPa)
Melting Pt:44…46 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:203-808-3
UN:2579
ADR:8,III
:GHS05
:GHS08
CAS Number:110-85-0
Formula:C₄H₁₀N₂
MDL Number:MFCD00005953