Barbituric acid
Supplier: Roth Carl
Jump to:
About this item
Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
- CAS Number:67-52-7
- Formula:C₄H₄N₂O₃
- MDL Number:MFCD00006666
- Molecular Weight:128.09 g/mol
- Merck Index:13,00966
- Boiling Pt:441 °C (1013 hPa)
- Melting Pt:250…252 °C
- Storage Temperature:Ambient
- EINECS:200-658-0
2 Options Available Below
Product Details & Documents
Synonyms: N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
- CAS Number:67-52-7
- Formula:C₄H₄N₂O₃
- MDL Number:MFCD00006666
- Molecular Weight:128.09 g/mol
- Merck Index:13,00966
- Boiling Pt:441 °C (1013 hPa)
- Melting Pt:250…252 °C
- Storage Temperature:Ambient
- EINECS:200-658-0



