Order Entry
Export
Orders LinkContactUsLinkComponent
Barbituric acid ≥99%

Barbituric acid ≥99%

Barbituric acid ≥99%
Certificates
Barbituric acid ≥99%
Barbituric acid ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 67-52-7 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Flash Point: 150°(302°F) pH: 2-3 Melting Point: ca 250° dec. Beilstein Reference: 120502 Merck Reference: 14,963 MDL No.: MFCD00006666

Synonyms:  N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
  • CAS Number:67-52-7
  • Formula:C₄H₄N₂O₃
  • MDL Number:MFCD00006666
  • Molecular Weight:128.09 g/mol
  • Merck Index:14,963
  • Boiling Pt:441 °C (1013 hPa)
  • Melting Pt:∼ 250 °C (dec.)
  • Flash Pt:150 °C (302 °F)
  • Storage Temperature:Ambient
  • EINECS:200-658-0
Product Family Options

Product Information