Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Diazabicyclo[2.2.2]octane
1,4-Diazabicyclo[2.2.2]octane ≥98%
1,4-Diazabicyclo[2.2.2]octane ≥98%
:
Thermo Fisher Scientific
1,4-Diazabicyclo[2.2.2]octane ≥98%
:
Thermo Fisher Scientific
:
280-57-9
:
:
C₆H₁₂N₂
:
MFCD00006689
:
112.17 g/mol
:
14,9669
:
174…176 °C
:
155…160 °C
:
1.140
:
62 °C (143 °F)
:
Ambient
:
205-999-9
:
1325
:
4.1,II
:
GHS02
:
GHS05
:
GHS07
msds_ALFAA14003
Thermo Scientific Acroseal 25 Years
Thermo Scientific Stockroom Essentials