


1,4-Diazabicyclo[2.2.2]octane 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:280-57-9
:
Formula:C₆H₁₂N₂
MDL Number:MFCD00006689
:112.17 g/mol
Merck Index:12,09801
Boiling Pt:174 °C (760 mmHg)
Melting Pt:155…160 °C
Density:1.14 g/cm³
Flash Pt:62 °C
Storage Temperature:Ambient
EINECS:205-999-9
UN:1325
ADR:4.1,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Water:≤2.0 % (K.F.)
- GC:≥96.0 %