Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexene

Cyclohexene

Glass bottle, 500 mL
Catalog # ROTH3451.1
Flammable
Health
Irritant
Environment
Supplier:   Roth Carl
Cyclohexene
Catalog # ROTH3451.1
Supplier:  Roth Carl
CAS Number:  110-83-8
Cyclohexene

Glass bottle, 500 mL

CAS Number:  110-83-8
Price
98,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • CAS Number:110-83-8
  • Formula:C₆H₁₀
  • MDL Number:MFCD00001539
  • Molecular Weight:82.15 g/mol
  • Merck Index:13,02756
  • Boiling Pt:83 °C (1013 hPa)
  • Melting Pt:–104 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • EINECS:203-807-8
  • UN:2256
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications