Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methylcyclohexane, Sigma-Aldrich®

Flammable
Health
Irritant
Environment
Supplier:   Merck
Methylcyclohexane, Sigma-Aldrich®
Supplier:  Merck
Certificates
Methylcyclohexane, Sigma-Aldrich®
Methylcyclohexane, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:108-87-2
  • Formula:C₆H₁₁CH₃
  • MDL Number:MFCD00001497
  • Molecular Weight:98.19 g/mol
  • Merck Index:13,06074
  • Boiling Pt:101 °C (1013 hPa)
  • Melting Pt:–126 °C
  • Density:0.7693 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • UN:2296
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-624-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 106,00 €
    106,00 €
    1 * 100 mL
       106,00 €

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 366,00 €
    366,00 €
    1 * 1 L
       366,00 €