1,1-Dimethylcyclohexane, Sigma-Aldrich®
Catalog # SIAL110493-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:112.22 g/mol
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–33.3 °C
Density:0.777 g/cm³ (20 °C)
EINECS:209-687-3
UN:2263
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:590-66-9
Formula:C₈H₁₆