Order Entry
Export
ContactUsLinkComponent
3-Methyl-1,2-cyclopentanedione
3-Methyl-1,2-cyclopentanedione
Catalog # MOLE67214708-100G
Supplier:  Molekula
CAS Number:  
3-Methyl-1,2-cyclopentanedione
Catalog # MOLE67214708-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :112.13 g/mol
Boiling Pt:210 °C (1013 hPa)
Storage Temperature:Ambient
EINECS:212-154-8
CAS Number:765-70-8
Formula:C₆H₈O₂
MDL Number:MFCD00001417