Order Entry
Export
ContactUsLinkComponent
1,1-Cyclopropanedicarboxylic acid, Sigma-Aldrich®
1,1-Cyclopropanedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL343412-5G
Supplier:  Merck
CAS Number:  
1,1-Cyclopropanedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL343412-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :130.1 g/mol
Boiling Pt:371 °C (1013 hPa)
Melting Pt:134…138 °C
ADR:8,III
 :M-Clarity™ quality level = MQ200
EINECS:209-917-2
UN:3261
CAS Number:598-10-7
Formula:C₅H₆O₄
MDL Number:MFCD00013727