
1,1-Cyclobutanedicarboxylic acid, Sigma-Aldrich®
Catalog # SIALC95803-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.13 g/mol
Boiling Pt:663 °C (1013 hPa)
Melting Pt:156…158 °C
Storage Temperature:Ambient
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:226-651-2
UN:3261
:GHS05
CAS Number:5445-51-2
Formula:C₆H₈O₄
MDL Number:MFCD00001325