Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexanecarbonyl chloride, Sigma-Aldrich®

Cyclohexanecarbonyl chloride, Sigma-Aldrich®

Supplier:   Merck
Cyclohexanecarbonyl chloride, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cyclohexanecarbonyl chloride, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride
  • CAS Number:2719-27-9
  • Formula:C₇H₁₁ClO
  • MDL Number:MFCD00001456
  • Molecular Weight:146.62 g/mol
  • Boiling Pt:184 °C (1013 hPa)
  • Melting Pt:29 °C
  • Density:1.08 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:220-322-7
  • UN:3265
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ100
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 27,90 €
    27,90 €
    1 * 5 g
       27,90 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 47,90 €
    47,90 €
    1 * 25 g
       47,90 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 80,30 €
    80,30 €
    1 * 100 g
       80,30 €