1,4-Cyclohexanedione, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:637-88-7
:
Formula:C₆H₈O₂
MDL Number:MFCD00001606
Boiling Pt:227 °C (1013 hPa)
Melting Pt:75…76 °C
Density:1.28 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:112.13 g/mol
Storage Temperature:Ambient
EINECS:211-306-0