Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Cyclohexanedione
1,2-Cyclohexanedione, Sigma-Aldrich®
1,2-Cyclohexanedione, Sigma-Aldrich®
1,2-Cyclohexanedione, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,2-Cyclohexanedione, Sigma-Aldrich®
Supplier:
Merck
Certificates
About this item
CAS Number:
765-87-7
:
Boiling Pt:
193 °C (1013 hPa)
Melting Pt:
33…35 °C
Density:
1.1305 g/cm³ (20 °C)
EINECS:
212-155-3
Related Information
Documentation
Certificates
Search by Lot Number
Documentation