Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Chlorocyclohexanone, Sigma-Aldrich®

Supplier:   Merck
2-Chlorocyclohexanone, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-Chlorocyclohexanone, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Boiling Pt:203 °C (1013 hPa)
  • Melting Pt:18…20 °C
  • Density:1.16 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:212-505-5
  • UN:3335
  • ADR:9,
  •  :M-Clarity™ quality level = MQ100
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 234,00 €
    234,00 €
    1 * 25 g
       234,00 €

  • Plastic bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 896,00 €
    896,00 €
    1 * 100 g
       896,00 €