Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Cyclooctadiene ≥97%

1,3-Cyclooctadiene ≥97%

Health
Flammable
Irritant
1,3-Cyclooctadiene ≥97%
Certificates
1,3-Cyclooctadiene ≥97%
1,3-Cyclooctadiene ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 1700-10-3 Molecular Formula: C8H12 Formula Weight: 108.18 Storage Temperature: 2°C to 8°C Flash Point: 24°(75°F) Refractive Index: 1.4930 Odour: Characteristic Melting Point: -53° Boiling Point: 142-144° Beilstein Reference: 1901033 UN No.: 2520 MDL No.: MFCD00001751

Synonyms:  Cycloocta-1,3-diene
  • CAS Number:1700-10-3
  • Formula:C₈H₁₂
  • MDL Number:MFCD00001751
  • Molecular Weight:108.18 g/mol
  • Boiling Pt:142…144 °C
  • Melting Pt:–53 °C
  • Density:0.869
  • Flash Pt:24 °C (75 °F)
  • Storage Temperature:Refrigerator
  • EINECS:216-929-1
  • UN:2520
  • ADR:3,III
Product Family Options

Product Information