Order Entry
Export
ContactUsLinkComponent
2,3-Dichloro-5,6-dicyano-p-benzoquinone
2,3-Dichloro-5,6-dicyano-p-benzoquinone
Catalog # MOLEM72374328
Supplier:  Molekula
CAS Number:  
2,3-Dichloro-5,6-dicyano-p-benzoquinone
Catalog # MOLEM72374328
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :227.01 g/mol
Merck Index:13,03088
Boiling Pt:396 °C (1013 hPa)
Melting Pt:209…214 °C
Density:1.713 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-542-2
UN:3439
ADR:6.1,III
CAS Number:84-58-2
Formula:C₈Cl₂N₂O₂
MDL Number:MFCD00001593