
2,3-Dichloro-5,6-dicyano-p-benzoquinone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84-58-2
:
Formula:C₈Cl₂N₂O₂
MDL Number:MFCD00001593
:227.01 g/mol
Merck Index:14,3063
Boiling Pt:396 °C (1013 hPa)
Melting Pt:214…218 °C
Density:1.713 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-542-2
UN:3439
ADR:6.1,III
:GHS06