
2,3-Dichloroquinoxaline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2213-63-0
:
Formula:C₈H₄Cl₂N₂
MDL Number:MFCD00006720
:199.04 g/mol
Boiling Pt:301 °C (1013 hPa)
Melting Pt:150…155 °C
EINECS:218-667-3
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Off-white to light brown Needles
- Infrared spectrum:Conforms
- Melting point:150 to 155 °C
- GC:≥96.0 %