
2,4-Quinolinediol, Sigma-Aldrich®
Catalog # SIALQ1336-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:161.16 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:159 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-711-0
:GHS07
CAS Number:86-95-3
:70254-43-2
Formula:C₉H₇NO₂
MDL Number:MFCD00006744