7-Hydroxycoumarin, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- CAS Number:93-35-6
- Formula:C₉H₆O₃
- MDL Number:MFCD00006878
- Molecular Weight:162.14 g/mol
- Merck Index:12,09981
- Boiling Pt:382 °C (1013 hPa)
- Melting Pt:204…206 °C
- Storage Temperature:Ambient
- EINECS:202-240-3
2 Options Available Below
Product Details & Documents
Synonyms: Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
- CAS Number:93-35-6
- Formula:C₉H₆O₃
- MDL Number:MFCD00006878
- Molecular Weight:162.14 g/mol
- Merck Index:12,09981
- Boiling Pt:382 °C (1013 hPa)
- Melting Pt:204…206 °C
- Storage Temperature:Ambient
- EINECS:202-240-3



