Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxycoumarin

7-Hydroxycoumarin

Supplier:   Roth Carl
7-Hydroxycoumarin
Supplier:  Roth Carl
Certificates
7-Hydroxycoumarin

2 Options Available Below

Product Details & Documents
Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162.14 g/mol
  • Merck Index:12,09981
  • Boiling Pt:382 °C (1013 hPa)
  • Melting Pt:204…206 °C
  • Storage Temperature:Ambient
  • EINECS:202-240-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 48,70 €
    48,70 €
    1 * 10 g
       48,70 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 342,00 €
    342,00 €
    1 * 100 g
       342,00 €