Order Entry
Export
Orders LinkContactUsLinkComponent
7-Hydroxycoumarin ≥98%

7-Hydroxycoumarin ≥98%

Irritant
7-Hydroxycoumarin ≥98%
Certificates
7-Hydroxycoumarin ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 93-35-6 Molecular Formula: C9H6O3 Formula Weight: 162.14 Melting Point: 228-232° Beilstein Reference: 127683 Merck Reference: 14,9847 MDL No.: MFCD00006878

Synonyms:  Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
  • CAS Number:93-35-6
  • Formula:C₉H₆O₃
  • MDL Number:MFCD00006878
  • Molecular Weight:162.14 g/mol
  • Merck Index:14,9847
  • Boiling Pt:382 °C (1013 hPa)
  • Melting Pt:228…232 °C
  • Storage Temperature:Ambient
  • EINECS:202-240-3
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 58,80 €
    58,80 €
    1 * 25 g
       58,80 €

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 173,00 €
    173,00 €
    1 * 100 g
       173,00 €