Chroman-4-one 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:491-37-2
:
Formula:C₉H₈O₂
MDL Number:MFCD00006840
:148.16 g/mol
Boiling Pt:127…128 °C (13 mmHg)
Melting Pt:34…40 °C
Density:1,1291 g/cm³ (99.8 °C)
Flash Pt:110 °C
EINECS:207-736-3
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals or low melting mass
- Infrared spectrum:Conforms
- Melting point:34 to 40 °C
- GC:≥95 %