2,2'-Dihydroxybiphenyl, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1806-29-7
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00002210
:186.21 g/mol
Boiling Pt:315 °C (1013 hPa)
Melting Pt:106…109 °C
Density:1.342 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:217-303-0