

2-Phenylaniline 97%, Sigma-Aldrich®
Catalog # A42409-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:169.23 g/mol
Boiling Pt:299 °C (lit.)
Melting Pt:47…50 °C (lit.)
Density:1.16 g/cm³ (21 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:153 °C (307.4 °F) (closed cup)
EINECS:201-990-9
:GHS08
:GHS07
Storage Temperature:Room Temperature
CAS Number:90-41-5
Formula:C₁₂H₁₁N
MDL Number:MFCD00007701