4,4'-Dichlorobiphenyl (PCB No. 15)
Catalog # EHERC20001500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:223.1 g/mol
EINECS:218-095-4
CAS Number:2050-68-2
Formula:C₁₂H₈Cl₂