2,2'-Dichlorobiphenyl (PCB No. 4)
Catalog # EHERC20000400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:223.1 g/mol
Boiling Pt:289 °C (1013 hPa)
Melting Pt:59…60 °C
CAS Number:13029-08-8
Formula:C₁₂H₈Cl₂