2,4',5-Trichlorobiphenyl (PCB No. 31)
Catalog # EHERC20003100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:257.55 g/mol
UN:3432
ADR:9,II
CAS Number:16606-02-3
Formula:C₁₂H₇Cl₃