3,3',4-Trichlorobiphenyl (PCB No. 35)
Catalog # PROOCIL-PCB-35-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:257.55 g/mol
UN:3432
ADR:9,II
CAS Number:37680-69-6
Formula:C₁₂H₇Cl₃