2,3',6-Trichlorobiphenyl (PCB No. 27)
Catalog # PROOCIL-PCB-27-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:257.55 g/mol
CAS Number:38444-76-7
Formula:C₁₂H₇Cl₃