Order Entry
Export
ContactUsLinkComponent
2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106)
2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106)
Catalog # PROOCIL-PCB-106-C
CAS Number:  
2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106)
Catalog # PROOCIL-PCB-106-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :326.44 g/mol
CAS Number:70424-69-0
Formula:C₁₂H₅Cl₅