2,3,3',4,5'-Pentachlorobiphenyl (PCB No. 108)
Catalog # EHERC20010800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:326.44 g/mol
CAS Number:70362-41-3
Formula:C₁₂H₅Cl₅