Order Entry
Export
ContactUsLinkComponent
2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)
2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)
Catalog # PROOCIL-PCB-105-C
CAS Number:  
2,3,3',4,4'-Pentachlorobiphenyl (PCB No. 105)
Catalog # PROOCIL-PCB-105-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :326.44 g/mol
UN:3432
ADR:9,II
CAS Number:32598-14-4
Formula:C₁₂H₅Cl₅