2,3',4'-Trichlorobiphenyl (PCB No. 33)
Catalog # EHERC20003300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:257.55 g/mol
CAS Number:38444-86-9
Formula:C₁₂H₇Cl₃