Order Entry
Export
ContactUsLinkComponent
 
2,2',6-Trichlorobiphenyl (PCB No. 19)
Catalog # PROOCIL-PCB-19-C
CAS Number:  
2,2',6-Trichlorobiphenyl (PCB No. 19)
Catalog # PROOCIL-PCB-19-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :257.55 g/mol
CAS Number:38444-73-4
Formula:C₁₂H₇Cl₃