2,2',5-Trichlorobiphenyl (PCB No. 18)
Catalog # EHERC20001800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:257.55 g/mol
Melting Pt:44 °C
Density:1.1485 g/cm³ (20 °C)
UN:3432
ADR:9,II
CAS Number:37680-65-2
Formula:C₁₂H₇Cl₃