Order Entry
Export
ContactUsLinkComponent
2,2',4,6-Tetrachlorobiphenyl (PCB No. 50)
2,2',4,6-Tetrachlorobiphenyl (PCB No. 50)
Catalog # PROOCIL-PCB-50-C
CAS Number:  
2,2',4,6-Tetrachlorobiphenyl (PCB No. 50)
Catalog # PROOCIL-PCB-50-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
UN:3432
ADR:9,II
CAS Number:62796-65-0
Formula:C₁₂H₆Cl₄