2,3',4',5,5'-Pentachlorobiphenyl (PCB No. 124)
Catalog # PROOCIL-PCB-124-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:326.44 g/mol
CAS Number:70424-70-3
Formula:C₁₂H₅Cl₅