2,2',4,5',6-Pentachlorobiphenyl (PCB No. 103)
Catalog # EHERC20010300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:326.44 g/mol
CAS Number:60145-21-3
Formula:C₁₂H₅Cl₅