2,2',3,3',4,5-Hexachlorobiphenyl (PCB No. 129)
Catalog # PROOCIL-PCB-129-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
CAS Number:55215-18-4
Formula:C₁₂H₄Cl₆