2,2',6,6'-Tetrachlorobiphenyl (PCB No. 54)
Catalog # EHERC20005400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
ADR:9,II
:291.99 g/mol
UN:3432
CAS Number:15968-05-5
Formula:C₁₂H₆Cl₄